Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse IL-1 alpha ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of protein levels from samples including serum, cell lysates, and cell culture supernatants. Interleukin- 1 alpha (IL-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells.

$332.05 - $941.05

Specifications

For Use With (Application) ELISA
Synonym Interleukin-1 alpha, IL1F1, Hematopoietin-1
Target IL-1 beta
Sensitivity 4pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125108

 
ebioscience
LS88501977
Each for $941.05

501125107

 
ebioscience
LS88501922
96-well plates 2 x 96 tests Each for $332.05

501125110

 
ebioscience
LS88501988
Each for $665.55

501125109

 
ebioscience
LS88501986
96-well plates 10 x 96 tests Each for $694.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym Interleukin-1 alpha, IL1F1, Hematopoietin-1
Target IL-1 beta
Sensitivity 4pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Mouse

Mouse IL-1 alpha Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)