Invitrogen™ eBioscience™ Mouse IL-1 alpha ELISA Ready-SET-Go!™ Kit
Specifically engineered for accurate and precise measurement of protein levels from samples including serum, cell lysates, and cell culture supernatants. Interleukin- 1 alpha (IL-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells.
Manufacturer:  Invitrogen™ LS88501977
 View more versions of this product
Catalog No. 501125108
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.
Description
Description
Mouse IL-1 alpha Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
ELISA
4pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Interleukin-1 alpha, IL1F1, Hematopoietin-1
20 x 96 tests
Mouse
IL-1 beta