Please login to your online account to display your discounted pricing

Mouse IgG2b Kappa Light Chain, Clone: MG2b, PE-Cy7, Isotype control, Novus Biologicals™

$252.00 - $373.00

Specifications

Antigen IgG2b Kappa Light Chain
Primary or Secondary Primary
Host Species Murine
Conjugate PE-Cy7
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14294201

 
novus biologicals canada
NBP14294201MG
0.1mg Each for $373.00

NBP14294205

 
novus biologicals canada
NBP142942005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen IgG2b Kappa Light Chain
Primary or Secondary Primary
Host Species Murine
Conjugate PE-Cy7
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.