missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IgG2a Kappa Light Chain, Clone: M2a, FITC, Isotype control, Novus Biologicals™

$219.00 - $329.00

Specifications

Antigen IgG2a Kappa Light Chain
Primary or Secondary Primary
Host Species Murine
Conjugate FITC
Applications Flow Cytometry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14395501 Novus Biologicals Canada
NBP14395501MG
0.1mg Each for $329.00
NBP14395505 Novus Biologicals Canada
NBP143955005MG
0.05mg Each for $219.00
Specifications Description & Specifications

Specifications

IgG2a Kappa Light Chain
Murine
Flow Cytometry
Primary
FITC
Store at 4°C in the dark.