Please login to your online account to display your discounted pricing

Mouse IgG2a Kappa Light Chain, Clone: eBM2a, Unlabeled, Isotype control, Novus Biologicals™

$175.00 - $307.00

Specifications

Antigen IgG2a Kappa Light Chain
Immunogen KLH and Freund's Adjuvant
Primary or Secondary Primary
Host Species Murine
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1433185

 
novus biologicals canada
NBP14331805MG
0.5mg Each for $307.00

NBP1433185M

 
novus biologicals canada
NBP143318005MG
0.05mg Each for $175.00
Description & Specifications

Specifications

Antigen IgG2a Kappa Light Chain
Immunogen KLH and Freund's Adjuvant
Primary or Secondary Primary
Host Species Murine
Conjugate Unlabeled
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.