Please login to your online account to display your discounted pricing

Mouse IgG2a Kappa Light Chain, Clone: eBM2a, Unlabeled, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143318005MG

 View more versions of this product

Catalog No. NBP1433185M

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Quantity 0.05mg
Host Species Mouse
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG2a Kappa
Clone eBM2a
Applications Flow Cytometry
Monoclonal or Polyclonal Monoclonal
Test Specificity This monoclonal mouse IgG2a, kappa immunoglobulin is used as an isotype control for mouse IgG2a antibodies.
Concentration 0.5mg/mL
Immunogen KLH and Freund's Adjuvant
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Antigen IgG2a Kappa Light Chain
Format Liquid