Please login to your online account to display your discounted pricing

Mouse IgG1 Kappa Light Chain, Clone: P3.6.2.8.1, Unlabeled, Isotype control, Novus Biologicals™

$175.00 - $307.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Murine
Conjugate Unlabeled
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1433195

 
novus biologicals canada
NBP14331905MG
0.5mg Each for $307.00

NBP1433195M

 
novus biologicals canada
NBP143319005MG
0.05mg Each for $175.00
Description & Specifications

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Murine
Conjugate Unlabeled
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blot
Storage Requirements Store at 4°C. Do not freeze.