Please login to your online account to display your discounted pricing

Mouse IgG1 Kappa Light Chain, Clone: P3.6.2.8.1, FITC, Isotype control, Novus Biologicals™

$219.00 - $329.00

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Mouse
Conjugate FITC
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14395601

 
novus biologicals canada
NBP14395601MG
0.1mg Each for $329.00

NBP14395605

 
novus biologicals canada
NBP143956005MG
0.05mg Each for $219.00
Description & Specifications

Specifications

Antigen IgG1 Kappa Light Chain
Primary or Secondary Primary
Host Species Mouse
Conjugate FITC
Applications Flow Cytometry
Storage Requirements Store at 4°C in the dark.