missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 506, eBioscience™, Invitrogen™

Mouse Isotype Control

$330.00 - $330.00

Specifications

Antigen Mouse IgG1 kappa
Clone P3.6.2.8.1
Concentration 0.2 mg/mL
Applications Control, Flow Cytometry
Conjugate eFluor 506
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124962
View Documents Promotion Details
Invitrogen
69471482
100 μg
Each for $330.00
 
Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: This P3.6.2.8.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.
Specifications

Specifications

Mouse IgG1 kappa
0.2 mg/mL
eFluor 506
Mouse
Not Applicable
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
P3.6.2.8.1
Control, Flow Cytometry
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
IgG1 κ
Affinity chromatography
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only.