Please login to your online account to display your discounted pricing

Mouse IgG1 Control

Mouse IgG1 Control from Thermo Scientific™ Pierce™ is validated for Ctrl, FACS and IHC (F).

$501.02 - $501.02

Products
Catalog Number Mfr. No. Price Quantity    

PIMA518168

 
pierce biotechnology inc
MA518168
Each for $501.02
Description & Specifications

Mouse IgG1 Control from Thermo Scientific™ Pierce™ is validated for Ctrl, FACS and IHC (F).

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody’s species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Host Species: Mouse
Isotype: IgG1
Immunogen: Rat thymocyte membrane glycoproteins
Formulation: 100 Tests of Protein A purified isotype control (0.1mg/mL) in PBS buffer, with BSA 1%;
MA5-18092 has been successfully used in Ctrl, FACS and IHC (F) procedures.