missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IFN-gamma Quantikine ELISA Kit, R&D Systems

$819.94 - $1,967.07

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 15978
Sample Type Cell Culture Supernates, Serum
Sample Volume Cell Culture Supernates (50μL), Serum (50μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MIF00 R & D Systems
RDSMIF00
2 Plates Each for $819.94
SMIF00 R & D Systems
RDSSMIF00
6 Plates Each for $1,967.07
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL)
Sensitivity: 2pg/mL
Specificity: Natural and recombinant mouse IFN-gamma
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
15978
Cell Culture Supernates (50μL), Serum (50μL)
Mouse
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum
2pg/mL
Natural and recombinant mouse IFN-gamma