missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Mouse IFN gamma ELISPOT ELISA Ready-SET-Go!™ Kit

This ELISPOT reagent set is pre-titrated for optimal spot development.

$785.40 - $785.40

Specifications

For Use With (Application) ELISPOT
Synonym Interferon gamma
Target IFN gamma
Storage Requirements 2° to 8°C
Species Mouse
View More Specs

Alert:

Contains Azide

Disclaimers:

For Research Use Only


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer This Ready-Set-Go! ELISPOT Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 uM).
5X ELISA/ELISPOT Diluent .
Enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501128959 Life Technologies
LS88738488
10 x 96 tests Each for $785.40
Specifications

Description

Mouse IFN gamma ELISPOT Ready-SET-Go! Reagent set contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IFNγ-secreting cells.
Description & Specifications

Specifications

ELISPOT
IFN gamma
Mouse
Interferon gamma
2° to 8°C