missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse IFN gamma ELISA Kits

Measure levels of mouse interferon gamma (IFNg) in mouse serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples using 96-well plates.

$651.05 - $2,436.00

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant mouse interferon gamma (IFNg)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
ENEM1001 Thermo Scientific™ Pierce™
PIEM1001
Mouse IFN gamma ELISA Kit 96 microplate wells Each for $651.05
ENEM10015 Thermo Scientific™ Pierce™
PIEM10015
Mouse IFN gamma ELISA Kit 5 x 96 microplate wells Each for $2,436.00
Specifications

Description

Assay IFN-γ (mouse interferon gamma) levels in mouse serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples with our Thermo Scientific Mouse IFN gamma ELISA Kits.

These mouse IFN-gamma ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interferon gamma (IFNγ) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IFNγ antibody pairs and IFNγ standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse interferon gamma (IFNγ)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 37 to 3000pg/mL

  • Sensitivity: < 10pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with with mouse IL-1α, IL-1β, IL-3, IL-4, IL-6, IL-7, IL-10, GM-CSF, TNFα or human IFNα or IFNγ

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

Description & Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV <10%; inter-assay CV <10%
Natural and recombinant mouse interferon gamma (IFNg)
Absorbance at 450nm minus absorbance at 550nm
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)