Learn More
Invitrogen™ IFN alpha Mouse ELISA Kit
For the quantitative detection of mouse IFN-alpha.
$782.00 - $5370.00
Specifications
Accession Number | P01572 |
---|---|
Assay Sensitivity | 7.48 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Sample Type | Plasma, Serum, Supernatant |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IFN-alpha
Biotin-Conjugate anti-mouse IFN-alpha monoclonal antibody
Streptavidin-HRP
Mouse IFN-alpha Standard lyophilized, 4000pg/mL upon reconstitution
Calibrator Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Mouse Interferon alpha (Ms IFNα) ELISA quantitates Ms IFNα in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IFNα. Principle of the method The Mouse IFNα solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
P01572 | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
15962 | |
7.2% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
Alpha interferon, IFN leukocytic, LeIF D | |
1 hr. 20 min. |
7.48 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
Ifa1 | |
3.7% | |
HRP | |
2°C to 8°C | |
Mouse | |
3 hr. 10 min. |
For Research Use Only