missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse IFN-alpha ELISA Kit, R&D Systems

to measure levels of IFN-α in tissue culture media

$655.69 - $2,762.24

Specifications

Sensitivity 12.5pg/mL
Specificity Detects all 14 Mouse IFN-a sub-types in tissue Culture Media, no-cross reactivity with Rat IFN-a, Mouse IFN-b, or Human IFN-a
Test Time 27.25 hr.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
421201 R & D Systems
RDS421201
96-well plate Each for $655.69
421202 R & D Systems
RDS421202
5 x 96-well plate Each for $2,762.24
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL)
Sensitivity: 12.5pg/mL
Specificity: Mouse IFN-α
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

12.5pg/mL
Detects all 14 Mouse IFN-a sub-types in tissue Culture Media, no-cross reactivity with Rat IFN-a, Mouse IFN-b, or Human IFN-a
27.25 hr.