Invitrogen™ eBioscience™ Mouse Granzyme B Platinum ELISA Kit
For the quantitative detection of mouse Granzyme B
Manufacturer:  Invitrogen™ LSBMS6029TEN
 View more versions of this product
Catalog No. 501125446
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse Granzyme B
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
Biotin
10 x 96 tests
Serum, plasma (citrate, EDTA), cell culture supernatant
Mouse
Granzyme 2, Fragmentin 2, Human lymphocyte protein
ELISA
1.6pg/mL
50μL
Granzyme B