Please login to your online account to display your discounted pricing

eBiosciencetrade; Mouse Granzyme B ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of Mouse Granzyme B protein levels in serum, plasma, and tissue culture supernant samples.

$332.05 - $694.55

Specifications

For Use With (Application) ELISA
Synonym GrzB, GzB
Target Granzyme B
Sensitivity 40pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
View More Specs
Includes:  Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

5017473

 
ebioscience&trade
LS88802222
Each for $332.05

5017474

 
ebioscience&trade
LS88802286
96-well plates 10 x 96 tests Each for $694.55

5017475

 
ebioscience&trade
LS88802288
Each for $665.55
Description & Specifications

Specifications

For Use With (Application) ELISA
Synonym GrzB, GzB
Target Granzyme B
Sensitivity 40pg/mL
Storage Requirements 2° to 8°C except standard which should be stored at ≤−70°C
Species Mouse

Mouse Granzyme B ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA).