Please login to your online account to display your discounted pricing

eBioscience™ Mouse GM-CSF, Recombinant Protein, Carrier-Free

Mouse Recombinant Protein

$1,115.05 - $2,739.05

Specifications

Name GM-CSF
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

509208

 
ebioscience™
LS34833182
100μg Each for $1,115.05

509209

 
ebioscience™
LS34833185
500μg Each for $2,739.05
Description & Specifications

Specifications

Name GM-CSF
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight 14kDa
Storage Requirements Store at less than or equal to -70°C.

Mouse Granulocyte/Macrophage Colony Stimulating Factor (GM-CSF) is an ∽14 kDa factor produced mainly by activated T cells and macrophages. Other cell types, such as endothelium and fibroblasts, also secrete GM-CSF in response to TNF alpha, IL-2, IL-1, and IFN gamma. GM-CSF stimulates growth of macrophages, granulocytes and dendritic cells. GM-CSF is found as a membrane-bound form and also as a complex associated with the extracellular matrix. Non-glycosylated GM-CSF is biologically active.