missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse GM-CSF ELISA Kits

Measure levels of granulocyte macrophage colony-stimulating factor in mouse serum and culture supernatants using 96-well plates.

$651.05 - $651.05

Specifications

Assay Range 0 to 250pg/mL
Description Mouse GM-CSF ELISA Kit
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Reproducibility Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
ENEMGMCSF Thermo Scientific™ Pierce™
PIEMGMCSF
96 microplate wells Each for $651.05
Specifications

Description

Assay GM-CSF (granulocyte macrophage colony-stimulating factor) levels in mouse serum and culture supernatants with our Thermo Scientific™ Mouse GM-CSF ELISA Kits.

These mouse GM-CSF ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of granulocyte macrophage colony-stimulating factor (GM-CSF) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse granulocyte-macrophage colony-stimulating factor (GM-CSF)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 0 to 250pg/mL

  • Sensitivity: < 5pg/mL

  • Reproducibility: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, IFNγ, TNFα, rabbit GM-CSF, rat GM-CSF or human GM-CSF

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Description & Specifications

Specifications

0 to 250pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Cell culture supernatant, serum
No significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, IFNg, TNFa, rabbit GM-CSF, rat GM-CSF or human GM-CSF
3.5 hr.
Mouse GM-CSF ELISA Kit
Absorbance at 450nm minus absorbance at 550nm
50µL
<5pg/mL
Natural and recombinant mouse granulocyte-macrophage colony-stimulating factor (GM-CSF)