Please login to your online account to display your discounted pricing

eBioscience™ Mouse Flt3 Ligand, Recombinant Protein

Mouse Recombinant Protein

$129.05 - $375.55

Specifications

Name Flt3 Ligand
Recombinant Yes
Conjugate Unlabeled
Purity >97%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

509310

 
life technologies
LS14800162
5μg Each for $129.05

509311

 
life technologies
LS14800180
25μg Each for $375.55
Description & Specifications

Specifications

Name Flt3 Ligand
Recombinant Yes
Conjugate Unlabeled
Purity >97%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight 20.7kDa
Storage Requirements Store at less than or equal to -70°C.

Flt3 (FMS-related tyrosine kinase 3) Ligand is a growth factor that stimulates the growth and differentiation of early hematopoetic progenitor cells. Flt3 Ligand alone exerts weak biological effects, and instead synergizes with other colony-stimulating factors to induce the proliferation and differentiation of cells of the myeloid and lymphoid lineages. Recent studies have suggested that Flt3 Ligand plays an important role in the development and expansion of dendritic cells in vivo, and that it may influence NK cell survival by this mechanism.

Flt3 Ligand transcripts are detectable in most cell types and tissues, although the protein has only been found in stromal cells and T cells. It is expressed most often as a transmembrane cell surface protein, although processing can yield a soluble, secreted isoform.