missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse CXCL2/MIP-2 Quantikine ELISA Kit, R&D Systems

a 4.5 hour solid phase ELISA designed to measure mouse MIP-2 levels in cell culture supernates and serum.

$819.94 - $1,967.07

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (50μL), Serum (50μL)
Species Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MM200 R & D Systems
RDSMM200
2 Plates Each for $819.94
SMM200 R & D Systems
RDSSMM200
6 Plates Each for $1,967.07
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL)
Sensitivity: 1.5pg/mL
Specificity: Natural and recombinant mouse MIP-2
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Solid Phase Sandwich ELISA
Cell Culture Supernates (50μL), Serum (50μL)
Mouse