Please login to your online account to display your discounted pricing

Mouse CXCL10/IP-10/CRG-2 Quantikine ELISA Kit, R&D Systems

Manufacturer: r & d systems  RDSMCX100

 View all options for this product

Catalog No. MCX100


Description & Specifications


Gene ID (Entrez) 15945
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 4.2pg/mL
Sample Type Cell Culture Supernates, Serum, Heparin Plasma
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), Heparin Plasma (50μL)
Species Mouse
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL), Heparin Plasma (50μL)
Sensitivity: 4.2pg/mL
Specificity: Natural and recombinant mouse IP-10
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.