Please login to your online account to display your discounted pricing

Mouse CXCL10/IP-10/CRG-2 Quantikine ELISA Kit, R&D Systems

$2,996.88 - $2,996.88

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), Heparin Plasma (50μL)
Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SMCX100

 
r & d systems
RDSSMCX100
6 Plates Each for $2,996.88
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), Heparin Plasma (50μL)
Species Mouse

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (50μL), Heparin Plasma (50μL)
Sensitivity: 4.2pg/mL
Specificity: Natural and recombinant mouse IP-10
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.