Please login to your online account to display your discounted pricing

Mouse CXCL10/IP-10/CRG-2 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSMCX100

 View more versions of this product

Catalog No. SMCX100

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 15945
Product Type Solid Phase Sandwich ELISA
Sensitivity 4.2pg/mL
Sample Volume Cell Culture Supernates (50μL), Serum (50μL), Heparin Plasma (50μL)
Species Mouse
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates
Packaging Quantity 6 Plates
Quantity 1 Pk.
Sample Type Cell Culture Supernates, Serum, Heparin Plasma
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.