Please login to your online account to display your discounted pricing

Mouse CXCL1/KC Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSMKC00B

 View more versions of this product

Catalog No. MKC00B

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
For Use With (Application) ELISA
Gene ID (Entrez) 14825
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 2 Plates
Quantity 1 Kit
Sensitivity 2pg/mL
Sample Type Cell Culture Supernates, Serum
Sample Volume Cell Culture Supernates (50μL), Serum (50μL)
Species Mouse
Sufficient For Sufficient materials to run ELISAs on at least two 96-well plates