Please login to your online account to display your discounted pricing

Mouse CCL5/RANTES DuoSet, R&D Systems

$828.14 - $828.14

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DY478

 
r & d systems
RDSDY478
15 96-well plates Each for $828.14
Description & Specifications

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Mouse

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.