Please login to your online account to display your discounted pricing

Mouse CCL3/MIP-1 alpha DuoSet, R&D Systems

$828.14 - $828.14

Specifications

Applications Development of sandwich ELISA to measure natural and recombinant mouse CCL3/MIP-1 alpha
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DY450

 
r & d systems
RDSDY450
15 x 96-well plate Each for $828.14
Description & Specifications

Specifications

Applications Development of sandwich ELISA to measure natural and recombinant mouse CCL3/MIP-1 alpha
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 Kit
Species Mouse

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.