Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: VWF635, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Von Willebrand Factor
Clone VWF635
Concentration 1mg/mL
Immunogen Recombinant human vWF fragment
Gene Accession No. P04275
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23451001

 
novus biologicals canada
NBP23451001MG
0.1mg Each for $384.00

NBP23451002

 
novus biologicals canada
NBP23451002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Von Willebrand Factor
Clone VWF635
Concentration 1mg/mL
Immunogen Recombinant human vWF fragment
Gene Accession No. P04275
Gene ID (Entrez) 7450
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot