Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: VWF635, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451002

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant human vWF fragment
Gene ID (Entrez) 7450
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Conjugate Unlabeled
Cross Reactivity Human
Antigen Von Willebrand Factor
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04275
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone VWF635
Concentration 1mg/mL