Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: SPM577, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Von Willebrand Factor
Clone SPM577
Concentration 0.2mg/mL
Immunogen Recombinant human vWF fragment
Gene Accession No. P04275
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23300401

 
novus biologicals canada
NBP23300401ML
0.1mL Each for $139.00

NBP23300410

 
novus biologicals canada
NBP23300410ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Von Willebrand Factor
Clone SPM577
Concentration 0.2mg/mL
Immunogen Recombinant human vWF fragment
Gene Accession No. P04275
Gene ID (Entrez) 7450
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting