Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: SPM577, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23300401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23300401

  • / Each
 

Description & Specifications

Specifications

Antigen Von Willebrand Factor
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Immunogen Recombinant human vWF fragment
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7450
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P04275
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone SPM577