Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: SPM577, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481002

  • / Each
 

Description & Specifications

Specifications

Antigen Von Willebrand Factor
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Recombinant human vWF fragment
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen.
Reconstitution Protein A purified
Clone SPM577
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P04275
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7450