Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: 3E2D10 + VWF635, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431005

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Clone 3E2D10 + VWF635
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Antigen Von Willebrand Factor
Immunogen Recombinant human vWF fragment (3E2D10 & VWF635)
Gene Accession No. P04275
Gene ID (Entrez) 7450
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Primary or Secondary Primary
Reconstitution Protein A purified
Conjugate Unlabeled
Quantity 0.5mL
Host Species Mouse
Cross Reactivity Human