Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: 3E2D10 + VWF635, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 7450
Cross Reactivity Human
Antigen Von Willebrand Factor
Formulation PBS with 0.05% BSA.
Gene Accession No. P04275
Immunogen Recombinant human vWF fragment (3E2D10 & VWF635)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 3E2D10 + VWF635
Conjugate Unlabeled
Regulatory Status RUO
Reconstitution Protein A purified
Concentration 0.2mg/mL