Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: 3E2D10 + VWF635, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453601

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Gene ID (Entrez) 7450
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Gene Accession No. P04275
Isotype IgG1, κ
Immunogen Recombinant human vWF fragment (3E2D10 & VWF635)
Regulatory Status RUO
Purification Method Purified
Quantity 0.1mg
Format Purified
Antigen Von Willebrand Factor
Clone 3E2D10 + VWF635
Concentration 1mg/mL
Primary or Secondary Primary
Reconstitution Protein A purified