Please login to your online account to display your discounted pricing

Mouse anti-Von Willebrand Factor, Clone: 3E2D10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453502

  • / Each
 

Description & Specifications

Specifications

Antigen Von Willebrand Factor
Cross Reactivity Human
Regulatory Status RUO
Quantity 0.2mg
Format Purified
Monoclonal or Polyclonal Monoclonal
Immunogen Recombinant human vWF fragment spanning aa 845-949
Clone 3E2D10
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Reconstitution Protein A purified
Gene Accession No. P04275
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Concentration 1mg/mL
Primary or Secondary Primary
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Gene ID (Entrez) 7450