Please login to your online account to display your discounted pricing

Mouse anti-Viperin, Clone: MaP.VIP, Novus Biologicals™

$332.66 - $586.39

Specifications

Antigen Viperin
Clone MaP.VIP
Concentration 0.1mg/mL
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Gene ID (Entrez) 91543
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14967701

 
novus biologicals canada
NBP14967701MG
0.1mg Each for $586.39

NBP14967705

 
novus biologicals canada
NBP149677005MG
0.05mg Each for $332.66
Description & Specifications

Specifications

Antigen Viperin
Clone MaP.VIP
Concentration 0.1mg/mL
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Gene ID (Entrez) 91543
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting