Please login to your online account to display your discounted pricing

Mouse anti-Viperin, Clone: MaP.VIP, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14967701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14967701

  • / Each
 

Description & Specifications

Specifications

Antigen Viperin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Clone MaP.VIP
Concentration 0.1mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin
Host Species Murine
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Isotype IgG2a
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 91543
Test Specificity Mouse Viperin
Target Species Human