Please login to your online account to display your discounted pricing

Mouse anti-Viperin, Clone: MaP.VIP, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149677005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14967705

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Host Species Mouse
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Concentration 0.1mg/mL
Primary or Secondary Primary
Test Specificity Mouse Viperin
Quantity 0.05mg
Gene ID (Entrez) 91543
Format Affinity Purified
Clone MaP.VIP
Monoclonal or Polyclonal Monoclonal
Purification Method Affinity Purified
Antigen Viperin
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Isotype IgG2a
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin