Please login to your online account to display your discounted pricing

Mouse anti-Viperin, Clone: MaP.VIP, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149603005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14960305

  • / Each
 

Description & Specifications

Specifications

Antigen Viperin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 91543
Format Affinity Purified
Formulation PBS, containing 0.1% BSA.
Host Species Mouse
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Test Specificity This antibody is specific for Viperin. Immunogen fragment used ranged from a.a. 92-362.
Clone MaP.VIP
Concentration 0.1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse