Please login to your online account to display your discounted pricing

Mouse anti-Viperin, Clone: MaP.VIP, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14960301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14960301

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, containing 0.1% BSA.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a
Primary or Secondary Primary
Gene ID (Entrez) 91543
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Viperin
Format Affinity Purified
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is specific for Viperin. Immunogen fragment used ranged from a.a. 92-362.
Clone MaP.VIP
Concentration 0.1mg/mL