Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: SPM576, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Vimentin
Clone SPM576
Concentration 0.2mg/mL
Immunogen Human vimentin recombinant protein
Gene Accession No. P08670
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23291001

 
novus biologicals canada
NBP23291001ML
0.1mL Each for $139.00

NBP23291010

 
novus biologicals canada
NBP23291010ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Vimentin
Clone SPM576
Concentration 0.2mg/mL
Immunogen Human vimentin recombinant protein
Gene Accession No. P08670
Gene ID (Entrez) 7431
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting