Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: SPM576, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23291010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23291010

  • / Each
 

Description & Specifications

Specifications

Antigen Vimentin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM576
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P08670
Host Species Mouse
Immunogen Human vimentin recombinant protein
Isotype IgG1
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7431
Test Specificity Does not react to Mouse and Rat.