Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: SPM576, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Vimentin
Clone SPM576
Concentration 1mg/mL
Immunogen Human vimentin recombinant protein
Gene Accession No. P08670
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23480901

 
novus biologicals canada
NBP23480901MG
0.1mg Each for $384.00

NBP23480902

 
novus biologicals canada
NBP23480902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Vimentin
Clone SPM576
Concentration 1mg/mL
Immunogen Human vimentin recombinant protein
Gene Accession No. P08670
Gene ID (Entrez) 7431
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting