Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: SPM576, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480902

  • / Each
 

Description & Specifications

Specifications

Antigen Vimentin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM576
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08670
Host Species Murine
Immunogen Human vimentin recombinant protein
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7431
Test Specificity This MAb reacts with a 58kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP's) such as desmin, keratin, neurofilament, and glial fibrillary acid protein. Does not react with Mouse
Reconstitution Protein A purified
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Feline
Target Species Goat
Target Species Human
Target Species Porcine