Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: SPM576, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Vimentin
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08670
Host Species Mouse
Immunogen Human vimentin recombinant protein
Isotype IgG1
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 7431
Clone SPM576
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb reacts with a 58kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP's) such as desmin, keratin, neurofilament, and glial fibrillary acid protein. Does not react with Mouse or Rat.
Reconstitution Protein A purified
Concentration 1mg/mL