Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: LN6, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Vimentin
Clone LN6
Immunogen Human thymic nuclear extract
Gene Accession No. P08670
Gene ID (Entrez) 7431
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23447001

 
novus biologicals canada
NBP23447001ML
0.1mL Each for $139.00

NBP23447005

 
novus biologicals canada
NBP23447005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Vimentin
Clone LN6
Immunogen Human thymic nuclear extract
Gene Accession No. P08670
Gene ID (Entrez) 7431
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Isotype IgM
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting