Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: LN6, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23447001ML

 View all options for this product


For Research Use Only

Catalog No. NBP23447001


Description & Specifications


Antigen Vimentin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone LN6
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P08670
Host Species Mouse
Immunogen Human thymic nuclear extract
Isotype IgM
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7431