Please login to your online account to display your discounted pricing

Mouse anti-Vimentin, Clone: LN6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23447005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23447005

  • / Each
 

Description & Specifications

Specifications

Antigen Vimentin
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P08670
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Human thymic nuclear extract
Isotype IgM
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7431
Clone LN6
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Host Species Mouse
Purification Method Concentrated
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.