Please login to your online account to display your discounted pricing

Mouse anti-VEGF R2/KDR/Flk-1, Clone: KDR721, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465801

  • / Each
 

Description & Specifications

Specifications

Antigen VEGF R2/KDR/Flk-1
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone KDR721
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P35968
Host Species Murine
Immunogen Recombinant human VEGF-R2 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3791
Reconstitution Protein A purified
Target Species Human